Order Entry
United States
ContactUsLinkComponent
Extech® Waterproof Palm pH Meter with Temperature
Extech® Waterproof Palm pH Meter with Temperature
Catalog # 470350-914
undefined
Extech® Waterproof Palm pH Meter with Temperature
Catalog # 470350-914
Supplier Number:  PH220-C

Specifications

  • Temperature Range:
    32° to 212°F (0 to 99.9°C)
  • Description:
    Waterproof pH meter with electrode
  • Temperature Accuracy:
    ±0.8°F/0.5°C
  • Temperature Resolution:
    0.1°
  • Compliance:
    IP57
  • Data Logging Capacity:
    25 readings
  • Dimensions:
    4.4 × 3.1 × 1.5" (111 × 79 × 39mm)
  • Display:
    LCD
  • Power supply:
    1 × 9V battery
  • Weight:
    9.2 oz (260 g)
  • pH Accuracy:
    ±0.01
  • pH Range:
    0.00 to 14.00
  • pH Resolution:
    0.01
  • Cat. No.:
    470350-914

Specifications

About this item

This palm-sized pH meter with cabled electrode can measure pH, voltage potential difference (mV) and temperature.

  • Internal memory stores up to 25 readings
  • Built-in microprocessor provides for automatic buffer recognition

This completely waterproof unit features a large LCD display with simultaneous measurement readings and low battery indicator.

Delivery: Includes cabled pH electrode, protective holster, pH buffers, and 9 V battery.