ContactUsLinkComponent
Gentamicin Sulfate, Reference Standard
Gentamicin Sulfate, Reference Standard
Catalog # 101095-074
Supplier:  USP
CAS Number:  
undefined
Gentamicin Sulfate, Reference Standard
Catalog # 101095-074
Supplier:  USP
Supplier Number:  1289003
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Pack type:
    Vial
  • CAS No.:
    1405-41-0
  • Description:
    Gentamicin Sulfate
  • Size:
    200 mg
  • Description:
    Gentamicin Sulfate
  • Cat. No.:
    101095-074
  • Certification:
    USP reference standard
  • Supplier no.:
    1289003
Specifications
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard