Orders LinkContactUsLinkComponent
Jailbreak Column Heater for Thermo Easy-Spray Source
Jailbreak Column Heater for Thermo Easy-Spray Source
Marketsource
Supplier:   PHOENIX S&T INC MS
Jailbreak Column Heater for Thermo Easy-Spray Source
Supplier:  PHOENIX S&T INC MS
Marketsource
Certificates
Jailbreak Column Heater for Thermo Easy-Spray Source
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Plug-and-Play directly into the Easy-Spray source and is supported by the trap-column bracket on the source.
  • Simple, flexible and low-cost
  • Temperature is controlled by the Easy-Spray source with the same precision as that of the OEM Easy-Spray columns.
The high voltage for nano spray comes from the source and is applied through a liquid junction - supply your own Tee or stainless steel union or purchase as a set.
Product Family Options
Product Information
  • Availability
    Price
  • MSPP-PSTESR17U
    Column heater for Easy-spray source, 17 cm
    Column heater for Easy-spray source, 17 cm
    Log in for availability
    Log in for availability
    Each
    Each - $4,441.54
    $4,441.54
    Each
       $4,441.54
    Supplier Number: PST-ESR-17U
    Specifications:
    Marketsource
    Marketsource
  • MSPP-PSTESR36U
    Column heater for Easy-spray source, 36 cm
    Column heater for Easy-spray source, 36 cm
    Log in for availability
    Log in for availability
    Each
    Each - $4,815.38
    $4,815.38
    Each
       $4,815.38
    Minimum order quantity is 1
    Supplier Number: PST-ESR-36U
    Specifications:
    Marketsource
    Marketsource