Order Entry
United States
ContactUsLinkComponent
Lens Cleaning Tissues, Radians
Lens Cleaning Tissues, Radians
Catalog # 82007-118
Supplier:  VisionAid, Inc.
Lens Cleaning Tissues, Radians
Catalog # 82007-118
Supplier:  VisionAid, Inc.
Supplier Number:  1LC1760

Specifications

  • Description:
    Lens Cleaning Tissues
  • L×W:
    7.5×5"
  • Cat. No.:
    82007-118
  • Supplier no.:
    1LC1760

Specifications

About this item

VisionAid Heavy Duty, Lint Free Tissues are stong and won't scratch lenses. Made of 100% recycled fiber.

  • High quality
  • Made of clean pulp, recyclable
Packaging: 600 Loose Tissues in a Pack.