Order Entry
United States
ContactUsLinkComponent
Thermoshaker for Microplates
Thermoshaker for Microplates
Catalog # MSPP-MB1004A
Supplier:  BIOTANG INC. MS
Irregular Voltage
Thermoshaker for Microplates
Catalog # MSPP-MB1004A
Supplier:  BIOTANG INC. MS
Supplier Number:  MB100-4A
Irregular Voltage

Specifications

  • Orbit:
    1.0 mm
  • Capacity:
    Up to 4 microplates
  • Description:
    Digital incubating microplate shaker
  • Tray dimensions:
    89W×133L×48H mm
  • Temperature Stability:
    ±0.3 °C
  • Heat-up Time:
    <25 min from 20...70 °C
  • Timer:
    0 - 99 hours 59 min (1 min increment)
  • Cat. No.:
    MSPP-MB1004A
  • W×D×H:
    320×350×185 mm
  • Electrical:
    110/220 V, 60/50 Hz
  • Speed range:
    100 - 1500 rpm
  • Weight:
    9 kg
  • Platform dimensions:
    133×89×48 mm (microplates)

Specifications

About this item

The MB100-4A is designed for shaking 1 to 4 standard 96-well plates in the thermostating mode.

  • Microprocessor controlled time, RPM, and temperature
  • Heating plates above and below the microplates
  • Incubation up to 70 °C in 0.1 °C steps
  • Shakes and incubates four microplates simultaneously
  • Beep-signal and stop of shaking motion after program completion
  • Excellent temperature accuracy and uniformity across the plate

It is designed using the multi-principle which allows using it as two independent devices:
1. Incubator for lasting incubation without shaking of micro-quantities (insect, plant cell cultures, etc.) in immunoplates.
2. Microplate thermo-shaker for immunochemistry and molecular diagnostics, where the requirements to the result reproducibility and thus to the precise method regulation are particularly high.

A distinctive feature for the MB100-4A is two-side heating of microtest plates that allows achieving exact correspondence of the set and actual temperature in wells of the microtest plates.

This device can be used in
1. Cytochemistry - for in situ reactions.
2. Immunochemistry - for immunofermentative reactions.
3. Biochemistry - for enzyme and protein analysis.
4. Molecular chemistry - for matrix analysis.