MonoSLEEVE™ Controller
Catalog # MSPP-MONOSLEEV
Supplier: ANALYTICAL SALES & SERVICE MS
Specifications
- Description:MonoSLEEVE single column heater controller
- Cat. No.:MSPP-MONOSLEEV
Specifications
About this item
MonoSLEEVE is the next generation of the original Hot Sleeve controller. With its new advanced “Plug & Play” technology, MonoSleeve can accommodate single HotSleeve+* or AgileSleeve+®* column heaters of different sizes without having to recalibrate to each individual sleeve. This allows you to switch out different heaters on the fly.
Delivery: Includes: MonoSLEEVE Controller, Power Supply, USB Cable, GUI, and Manual. Heater not included.
Caution: The temperature controller must be operated away from liquids so as not to accidentally spill solvents on the top cover. Do not immerse or operate any part of the Column Heater in liquids. In the event of solvent leakage, wipe sleeve clean before further use.