Order Entry
United States
Orders LinkContactUsLinkComponent
 

Linearity FD D-Dimer for Stago Systems, AUDIT Microcontrols

Linearity FD D-Dimer for Stago Systems, Freeze Dried, D-Dimer, 5x1 ml
Catalog # 76717-394
Supplier:   Audit Microcontrols
 :  K958M-5
Linearity FD D-Dimer for Stago Systems, AUDIT Microcontrols
Catalog # 76717-394
CAS Number:  
Linearity FD D-Dimer for Stago Systems, AUDIT Microcontrols
Linearity FD D-Dimer for Stago Systems, AUDIT Microcontrols

Linearity FD D-Dimer for Stago Systems, Freeze Dried, D-Dimer, 5x1 ml

CAS Number:  
Price
$421.35
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Linearity FD D-Dimer for Stago Systems is assayed quality control material consisting of five levels of human plasma.

  • Each level contains the following analytes: D-Dimer
  • These five levels demonstrate a linear relationship to each other for their respective analytes
  • It is intended to simulate human patient plasma samples for purpose of determining linearity, calibration verification and verification of reportable range for D-Dimer.
Specifications

Specifications

  • Description:
    Linearity FD D-Dimer for Stago Systems
  • Size:
    5x1 ml
  • Analyte:
    D-Dimer
  • Type:
    Freeze Dried

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.