G-TRAP™ Empty FPLC Columns
About this item
These empty columns are designed for fast protein liquid chromatography.
- Frit material is ultra-high molecular weight polyethylene
Columns are made of polypropylene and come in 1 ml and 5 ml sizes.
Delivery information: Includes column tube, sealing plug, sealing sleeve, top stop plug and bottom stop plug, top (large) frit, bottom (small) frit (inserted in column).
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
These empty columns are designed for fast protein liquid chromatography.
- Frit material is ultra-high molecular weight polyethylene
Columns are made of polypropylene and come in 1 ml and 5 ml sizes.
Delivery information: Includes column tube, sealing plug, sealing sleeve, top stop plug and bottom stop plug, top (large) frit, bottom (small) frit (inserted in column).
Specifications for Product Family
Specifications
- Catalog No:
- 76263-822
- 76263-824
- Max. Pressure:
- 3 bar
- 3 bar
Specifications
Product Family Options
Product Information
- Column hardwareCapacityAvailabilityPrice
- Supplier Number: 786-1290Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76263-822Max. Pressure3 bar - Supplier Number: 786-1291Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76263-824Max. Pressure3 bar