Anti-HSP90AA1/HSP90AB1 Mouse Monoclonal Antibody [clone: MBH90AB]
Catalog # 10673-568
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Symbol:HSP90AA1/B1
- Clonality:Monoclonal
- Clone:MBH90AB
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Bovine,Mouse
- Western Blot:Yes
- Size:100 μg
- Epitope:N-terminal
- Cross Adsorption:No
- Format:Liquid
- Storage Buffer:PBS (0.2% BSA, 15 mM sodium azide)
- Sequence:EEVHHGEEEVEC
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids at N-terminus of human HSP90AA1/HSP90AB1.
- Cat. No.:10673-568
- Supplier no.:MAB6457
Specifications
About this item
Mouse monoclonal antibody raised against synthetic peptide of HSP90AA1 and HSP90AB1.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: HSP90AA1/HSP90AB1
Clonality: Monoclonal
Clone: MBH90AB
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse