Anti-Static Circuit Chiller & Freeze Spray, MicroCare
Catalog # 76019-930
Supplier: MicroCare
Specifications
- Description:Anti-Static Circuit Chiller & Freeze Spray
- Size:10 oz.
- Cat. No.:76019-930
Specifications
About this item
This anti-static circuit chiller freeze spray quickly finds circuit board faults.
- No residue / no rinsing
- Leaves ice on the PCB momentarily to freeze it
- Dries instantly
- Chills to lower than -45˚F/-43ºC in seconds; may go as low as -60˚F/-51˚C depending upon ambient conditions
- Safe on plastics
- Chemical Family: HFC blend
- Nonflammable for better safety
This ultra-pure anti-static circuit chiller freeze spray uses an HFC propellant. So, it will not generate the component-shattering thermal shock that can occur with old-style HCFC-based chillers. ESD-safe when used in combination with the StatZap™ ESD Eliminator Tool.
Applications:
- Use for circuit test and repair
- Works on old and new equipment
Certifications: RoHS/WEEE/REACH/EPA SNAP compliant