Order Entry
United States
ContactUsLinkComponent
Multiparameter PH/EC/DO Turbidity Meter
Multiparameter PH/EC/DO Turbidity Meter
Catalog # 470349-824
Supplier:  HANNA INSTRUMENTS
undefined
Multiparameter PH/EC/DO Turbidity Meter
Catalog # 470349-824
Supplier:  HANNA INSTRUMENTS
Supplier Number:  HI9829-01041

Specifications

  • Calibration:
    pH: Automatic at 1, 2 or 3 points with automatic recognition of 5 standard buffers (pH 4.01, 6.86, 7.01, 9.18, 10.01) and 1 custom buffer
    ORP: Automatic at 1 custom point (relative mV)
    Conductivity: Automatic 1 point, with 6 standard solutions (84 µS/cm, 1413 µS/cm, 5.00 mS/cm, 12.88 mS/cm, 80.0 mS/cm, 111.8 mS/cm) or custom point
    Total Dissolved Solids (TDS): Based on conductivity calibration
    Dissolved Oxygen: Automatic 1 or 2 points at 0, 100% or 1 custom point
    Turbidity: Automatic 1, 2 or 3 points at 0, 20 and 200 FNU or custom
  • Description:
    Multiparameter PH/EC/DO turbidity meter
  • Turbidity - Range:
    0 - 99.9 FNU, 100 - 1000 FNU
  • Turbidity - Resolution:
    0.0 - 99.9 FNU, 100 - 1000 FNU
  • Turbidity - Accuracy:
    ±0.3 FNU or ±2% of reading, whichever is greater
  • Operating Temperature Range:
    0...50 °C (32...122 °F), RH 100%
  • DO Range:
    0.0 - 500.0%, 0.00 - 50.00 ppm (mg/L)
  • DO Resolution:
    0.1%, 0.01 ppm (mg/L)
  • Conductivity Accuracy:
    ±1% of reading or ±1 µS/cm whichever is greater
  • Conductivity Range:
    0 - 200 mS/cm (absolute EC up to 400 mS/cm)
  • Conductivity Resolution:
    Manual: 1 µS/cm, 0.001 mS/cm, 0.01 mS/cm, 0.1 mS/cm, 1 mS/cm
    Automatic: 1 µS/cm from 0 - 9999 µS/cm, 0.01 mS/cm from 10.00 - 99.99 mS/cm, 0.1 mS/cm from 100.0 - 400.0 mS/cm
    Automatic (mS/cm): 0.001 mS/cm from 0.000 - 9.999 mS/cm, 0.01 mS/cm from 10.00 - 99.99 mS/cm, 0.1 mS/cm from 100.0 - 400.0 mS/cm
  • DO Accuracy:
    0.0 - 300.0%: ±1.5% of reading or ±1.0% whichever is greater
    300.0 - 500.0%: ±3% of reading; 0.00 - 30.00 ppm (mg/L): ±1.5% of reading or ±0.10 ppm (mg/L) whichever is greater
    30.00 ppm (mg/L) - 50.00 ppm (mg/L): ±3% of reading
  • TDS Accuracy:
    ±1% of reading or ±1 ppm (mg/L) whichever is greater
  • TDS Range:
    0 - 400000 ppm (mg/L) (the maximum value depends on the TDS factor)
  • TDS Resolution:
    Manual: 1 ppm (mg/L), 0.001 ppt (g/L), 0.01 ppt (g/L), 0.1 ppt (g/L), 1 ppt (g/L)
    Automatic: 1 ppm (mg/L) from 0 - 9999 ppm (mg/L), 0.01 ppt (g/L) from 10.00 - 99.99 ppt (g/L), 0.1 ppt (g/L) from 100.0 - 400.0 ppt (g/L)
    Automatic ppt (g/L): 0.001 ppt (g/L) from 0.000 - 9.999 ppt (g/L), 0.01 ppt (g/L) from 10.00 - 99.99 ppt (g/L), 0.1 ppt (g/L) from 100.0 - 400.0 ppt (g/L)
  • mV accuracy:
    ±1.0 mV (ORP)
  • mV range:
    ±2000.0 mV (ORP)
  • mV resolution:
    0.1 mV (ORP)
  • pH Accuracy:
    ±0.02 pH
  • pH Range:
    0.00 - 14.00 pH
  • pH Resolution:
    0.01 pH
  • Cat. No.:
    470349-824

Specifications

About this item

This multiparameter meter is a waterproof portable logging multiparameter meter. The microprocessor based multi-sensor probe allows for the measurement of key parameters including pH, ORP, conductivity, dissolved oxygen, turbidity, and temperature. The complete system is simple to setup and easy to use.

  • Color coded, field replaceable sensors
  • Great for environmental, field, and laboratory testing
  • Digital probe that is less than 2" in diameter
  • 3 color-coded sensor inputs
  • IP 68 rating for continuous immersion in water
  • Multistrand - multiconductor cable rated for 150 lbs. (intermittent use)
  • Weighted shield protects all sensors
  • Quick calibration - The quick calibration feature allows for a single point calibration for pH, conductivity, and dissolved oxygen all in few simple steps

Waterproof Protection - The water quality meter is enclosed in an IP 67 rated waterproof casing and can withstand immersion in water at a depth of 1 m for up to 30 minutes. The probe features an IP68 rating for continuous immersion in water Backlit Dot Matrix LCD Display - The meter features a backlit graphic LCD with on-screen help and the capability to display up to twelve parameters simultaneously. The graphic display allows for the use of virtual keys to provide for an intuitive user interface.

Intuitive Keypad - The fitted rubber keypad has dedicated keys for power, backlight, up/down arrows, help and alphanumeric characters. The meter also features two virtual soft keys that navigate the user through the configuration of each parameter, meter setup and logging of data. The interface is intuitive for any user’s level of experience. Dedicated Help Key - Contextual help is always available through a dedicated ‘HELP’ key. Clear tutorial messages and directions are available on-screen to guide users quickly and easily through setup and calibration. The help information displayed is relative to the setting/option being viewed.

Auto-sensor Recognition - The probe and meter automatically recognize the sensors that are connected. Any ports not used on the probe will not have the parameter displayed or be configurable
Automatic temperature compensation - Integrated temperature sensor allows for automatic temperature compensation of pH, conductivity, and dissolved oxygen measurements. Quick Calibration - Quick Calibration provides a speedy, single point calibration for pH, conductivity, and dissolved oxygen. Standard calibration options are available including pH up to three points, Conductivity at one point and dissolved oxygen up to two points.

Data Logging - This meter allows users to store up to 44,000 continuous or log-on-demand samples with logging intervals from one second to three hours. Rugged thermoformed carrying case - The HI9829 meter, probe, and all accessories are supplied in a rugged carrying case designed to provide years of use. The inside compartment of the carrying case is thermoformed to securely hold and protect all the components.

Ordering information: Replacement probes and sensors sold seperately. Calibration solutions sold seperately. pH storage and cleaning solutions sold seperately. Contact your sales representative for more information.