Order Entry
United States
ContactUsLinkComponent
 
High Range PH/EC/TDS Tester
  470349-836
undefined
High Range PH/EC/TDS Tester
  470349-836
 :  HI98130

 

  • Description:
    High range pH/EC/TDS tester
  • Operating Temperature Range:
    0...50 °C (32...122 °F) [temperature of sample]
  • Conductivity Accuracy:
    ±2% F.S.
  • Conductivity Range:
    0.00 - 20.00 mS/cm
  • Conductivity Resolution:
    0.01 mS/cm
  • TDS Accuracy:
    ±2% F.S.
  • TDS Calibration:
    Automatic, one point at 12.88 mS/cm or 6.44 g/L (ppt)
  • TDS Range:
    0.00 - 10.00 g/L (ppt)
  • TDS Resolution:
    0.01 g/L (ppt)
  • pH Accuracy:
    ±0.05 pH
  • pH Calibration:
    Automatic, one or two-point with two sets of standard buffers (pH 4.01/7.01/10.01 or 4.01/6.86/9.18)
  • pH Range:
    0.00 - 14.00
  • pH Resolution:
    0.01 pH
  • Cat. No.:
    470349-836

 

 

Accurate. Waterproof. No more switching between meters for your routine measurements with the HI98130 pH and conductivity tester.

  • Compact and waterproof design fits right in your pocket so you can take it anywhere
  • Extractable cloth junction extends the life of your pH and conductivity tester
  • Great for hydroponics, aquariums, pools, and ponds
  • 3 sensors in a single meter
  • pH electrode with extractable junction
  • Amperometry EC/TDS sensor
  • Exposed temperature probe for faster EC/TDS readings

This multiparameter tester is a highly accurate, waterproof combo pH EC tester that can also measure EC/TDS in the high range (0.00 to 20.00 mS/cm EC/ 0.00 to 10.00 ppt TDS) and temperature. This floating, waterproof combination tester has an easy to read, LCD and an automatic shut-off. In addition, pH and EC/TDS readings are automatically temperature compensated (ATC) to prevent temperature-related variations in your measurements. For EC/TDS readings, the EC/TDS conversion factor (CONV) is selectable by the user, as well as the temperature compensation coefficient β (BETA).

 : Replaceable pH electrode is purchased seperately. pH calibration solutions, storage solutions, and cleaning solutions need to be purchased seperately. Contact your sales representative for more information.