ContactUsLinkComponent
Waterproof Pocket pH Tester
Waterproof Pocket pH Tester
  470349-826
undefined
Waterproof Pocket pH Tester
  470349-826
 :  HI98107U
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Waterproof pocket pH tester
  • pH Accuracy:
    ±0.2
  • pH Calibration:
    Automatic, two-points
  • pH Range:
    0.00 - 14.00
  • pH Resolution:
    0.1
  • Cat. No.:
    470349-826
This pH tester is an easy-to-use pH water tester that makes monitoring water quality simple. This accurate pH tester features 0.1 resolution with automatic two-point calibration and temperature compensation in a single, portable, pocket device.
  • Compact design fits right in your pocket so you can take it anywhere
  • Cloth junction providing a larger surface area than a standard ceramic junction
  • Great for drinking water, aquariums, pools, and ponds
Features a large multi-level LCD which displays both pH and temperature readings simultaneously. The pH readings are displayed with a 0.1 pH resolution and with an accuracy of ±0.1 pH. The pH range of the HI98107 is from 0.0 to 14.0 pH. An exposed temperature sensor allows for rapid automatic temperature compensated pH measurements. Temperature can be set to display in ℃ or ℉. The LCD screen has stability and calibration tag indicators. The battery percent level is displayed at start up alerting the user to the remaining battery power that is available.
The pH electrode uses a unique reference cell design that utilizes a cloth junction that provides a larger surface area than a standard ceramic junction.
Calibration is performed by pressing the dedicated calibration button. Once in calibration mode the pH electrode can be calibrated to 1 or 2 points using the pre-programmed standard buffers (pH 4.01, 7.01, 10.01). When the standard value is recognized and stability is reached, the meter automatically accepts and stores the calibration value.
Setup options include automatic shut off timing and temperature unit displayed. Automatic shut-off options are to turn the water pH meter off after 8 minutes, 60 minutes, or disabled. To enter the setup mode the user presses a button located in the battery compartment.
 : If pH probe dies, will have to repurchase product. pH calibration solutions, storage solutions, and cleaning solutions sold seperately. Contact your sales representative for more information.