- Description:HALO2 Bluetooth® pH tester, lab testing
- pH Accuracy:±0.02
- pH Calibration:Up to three points or four points
*Automatic buffer recognition with standard buffers Hanna (pH 1.68 *, 4.01, 7.01, 10.01) or NIST (pH 1.68 *, 4.01, 6.86, 9.18) - pH Range:0.00 - 12.00
- pH Resolution:0.01 or 0.1
- Cat. No.:470349-828
The HALO2 Wireless General Use pH Tester has a refillable glass body and spherical glass sensing bulb made for users that prefer a low-maintenance laboratory-grade pH electrode.
- Open-source Bluetooth allows easy integration into current data logging systems
- IP65 water resistant protection
- Specialized electrode for small volumes
Cleaning and disinfection solution – Your HALO2 pH tester comes with cleaning solution that has been engineered to keep your electrode free from grease and debris. This concentrated combination of detergents cleans your meter quickly and effectively so that your meter stays in optimum condition.
Easy calibration – HALO2 can be placed right into our calibration buffer sachets for easy calibration. Using a Hanna calibration sachet ensures your buffer is always fresh.
Storage solution – Proper storage of your electrode will ensure reliable measurements and a long electrode life. Our storage solution is specially formulated to keep the pH glass hydrated while minimizing organic growth.