Order Entry
United States
ContactUsLinkComponent
Free and Total Chlorine Portable Photometer
Free and Total Chlorine Portable Photometer
Catalog # 470349-850
Supplier:  HANNA INSTRUMENTS
undefined
Free and Total Chlorine Portable Photometer
Catalog # 470349-850
Supplier:  HANNA INSTRUMENTS
Supplier Number:  HI97711C

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Photometer kit, free total chlorine, portable
  • Model:
    HI97711
  • ISE Accuracy:
    ±0.03 mg/L, ±3% of reading at 25 °C
  • W×D×H:
    142.5 × 102.5 × 50.5 mm (5.6 x 4.0 x 2.0")
  • Batteries:
    1.5V AA battery (3 pcs.) / > 800 measurements (without backlight)
  • Data Logging - Log Function:
    Automatic (50 measurements)
  • Display:
    Backlit LCD
  • ISE Resolution:
    0.01 mg/L
  • ISE Range:
    0.00 - 5.00 mg/L
  • Weight:
    380 g (13.4 oz.) with batteries
  • Cat. No.:
    470349-850
  • Range:
    0.00 - 5.00 mg/L

Specifications

About this item

The Free and Total Chlorine Photometer combines accuracy and ease of use in a simple, portable design. The advanced optical system provides lab-quality accuracy while its user-friendly design is easy for any user making it the perfect photometer for your water quality testing needs. This meter measures free and total chlorine in water samples from 0.00 - 5.00 mg/L (ppm).

  • No warmup time needed before taking a measurement
  • Tutorial mode for easy step-by-step instructions
  • Adaptation of the EPA DPD method 330.5

Better features for better testing - Let your photometer guide you through setup, calibration, and testing with on-screen tutorials and animations so you always know the next step. Simply access Tutorial Mode at any stage in the setup or measurement process with the dedicated help key. The auto-off feature prolongs battery life for peace-of-mind and energy savings if the meter is accidentally left on. When the battery level is below 10%, a warning symbol will blink to indicate low battery condition. When low battery power may affect your results, the meter will shut down to avoid erroneous readings caused by low battery level.

Portable design - Made to fit comfortably in your hand for on-the-go testing or sit on a table for benchtop use, a backlit LCD display provides at-a-glance readings from any angle, day or night. The supplied AA batteries provide life for more than 800 measurements.

Ordering information: Reagents need to be purchased separately. Contact your sales representative for more information.
Delivery: Kit is supplied with CAL-Check standards, cuvettes (2), caps (2), plastic inserts for cuvettes (2), scissors, cuvette cleaning cloth, 1.5V AA batteries (3), instruction manual, and instrument quality certificate.