NEBExpress® T4 Lysozyme
Supplier: New England Biolabs (NEB)
Certificates
About this item
NEBExpress T4 Lysozyme is 200-fold more active than chicken egg white lysozyme.
- Lysis reactions are scalable and compatible with high throughput workflows
- Fast non-mechanical bacterial lysis; the lysate is ready to use and compatible with affinity resins
- Small (200 µg) and large (5×200 µg) formats suitable for lysing 7 or 35 g of cells, respectively
- Recombinant, animal free and REACH compliant
NEBExpress T4 Lysozyme is a recombinant murein hydrolase that breaks down the bacterial cell wall by hydrolyzing the beta 1 to 4 linkage between N-acetylmuramic acid and N-acetylglucosamine in the peptidoglycan of prokaryotic cells (gram-negative and some gram-positive bacteria). It can be used to extract soluble proteins, membrane proteins, DNA, RNA, or metabolites.
Frequently Bought Together