Human Follistatin(FS) ELISA Kit
76699-088
:
- Assay Type:Sandwich
- Host:Human
- Target protein:Follistatin
- Description:Human Follistatin(FS) ELISA Kit
- Size:96 wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.8 - 40 ng/ml
- Storage Temperature:2…8 °C
- Application:Signal Transduction
- Sample Volume:50 - 100 µl
- Sensitivity:0.1 ng/ml
- Regulatory Status:RUO
- Cat. No.:76699-088
- Detection wavelength: 450 nm