Vernier Go Direct® Sensor Clamp
470320-000
- Description:Vernier Go Direct® sensor clamp
- Cat. No.:470320-000
Secure your Go Direct® sensors for field work.
- Clamps securely to wand-style sensors
- Includes lanyard
The Go Direct® Sensor Clamp securely fastens to a wand-style Go Direct® sensor, and the included lanyard works as a strap to prevent accidental drops during investigations in the field.