Order Entry
United States
ContactUsLinkComponent
 
Human Insulin-like Growth Factor 2 mRNA-binding Protein 3(IGF2BP3) ELISA Kit
Catalog # 76708-368
Supplier:  AFG Bioscience
CAS Number:  
undefined
Human Insulin-like Growth Factor 2 mRNA-binding Protein 3(IGF2BP3) ELISA Kit
Catalog # 76708-368
Supplier:  AFG Bioscience
Supplier Number:  EK710178
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Host:
    Human
  • Target protein:
    Insulin-like growth Factor 2 mRNA-binding protein 3
  • Description:
    Human Insulin-like growth Factor 2 mRNA-binding protein 3(IGF2BP3) ELISA Kit
  • Size:
    96 wells
  • Sample Type:
    Serum, plasma, urine, tissue homogenates, cell culture supernates
  • Time to Results:
    1 - 5 h
  • Assay Principle:
    Quantitative
  • Shelf Life:
    6 months (4 °C); 12 months (–20 °C)
  • Detection Range:
    37.5 - 2400 pg/ml
  • Storage Temperature:
    2…8 °C
  • Application:
    Epigenetics and Nuclear Signaling, Signal Transduction
  • Sample Volume:
    50 - 100 µl
  • Sensitivity:
    100 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76708-368

Specifications

About this item

  • Detection wavelength: 450 nm