Vernier Physics LabQuest® Mini Standard Package
470225-078
- Description:Vernier Physics LabQuest® mini standard package
- Cat. No.:470225-078
- Supplier no.:LM-PHY-DX
Includes interface and suite of sensors for a lab group of 2 to 4 students.
- Sensors have wired connections, with BTA plugs
- LabQuest® Mini provides a convenient interface with a PC, Mac, or Chromebook
- 10 sensors and 2 accessories
- LabQuest® Mini included
This standard package is perfect for physics teachers setting up their probeware kit. The standard package comes with most of the sensors needed for on-level physics (mechanics and E&M), and supports one lab group of 2 to 4 students.
: Standard package includes LabQuest Mini (Model 2), Motion Detector, Dual-Range Force Sensor, Microphone, Differential Voltage Probe (2), Low-g Accelerometer, Light Sensor, Photogate (2), Ultra Pulley Attachment, Picket Fence, Stainless Steel, Temperature Probe, Magnetic Field Sensor, Current Probe (2).