ContactUsLinkComponent
Hand-Held Weather Center
Hand-Held Weather Center
  470332-742
undefined
Hand-Held Weather Center
  470332-742
 :  LER2940
  • Description:
    Weather center box
  • Cat. No.:
    470332-742
Easily carry around this economical weather station!
This set of three weather-measuring instruments includes its own carrying case and includes a 3' barometer (for measuring atmospheric pressure), hygrometer (for measuring humidity), and a Celsius and Fahrenheit thermometer. Individual stands are also included in the case. A helpful guide explains how to use each instrument.