Human Probable Prolyl tRNA Synthetase, Mitochondrial (PARS2) ELISA Kit
Catalog # 76705-096
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:Probable prolyl-tRNA synthetase, mitochondrial
- Description:Human Probable prolyl-tRNA synthetase, mitochondrial(PARS2) ELISA Kit
- Size:96 wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf Life:6 months (4 °C); 12 months (−20 °C)
- Storage Temperature:2…8 °C
- Application:Epigenetics and nuclear signaling
- Sample Volume:50 - 100 µl
- Sensitivity:6.25 pg/ml
- Regulatory Status:RUO
- Cat. No.:76705-096
Specifications
About this item
- Detection wavelength: 450 nm