Human Antibacterial Peptide LL 37 (APLL 37) ELISA Kit
Catalog # 76712-022
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:Antibacterial Peptide LL-37
- Description:Human Antibacterial Peptide LL-37 (APLL-37) ELISA Kit
- Size:96 wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf Life:6 months (4 °C); 12 months (−20 °C)
- Detection Range:0.375 - 12 ng/ml
- Storage Temperature:2…8 °C
- Sample Volume:50 - 100 µl
- Sensitivity:0.039 ng/ml
- Regulatory Status:RUO
- Cat. No.:76712-022
Specifications
About this item
- Detection wavelength: 450 nm