Human Nerve Growth Factor Receptor (NGFR) ELISA Kit
76715-962
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Assay Type:Sandwich
- Host:Human
- Primary antibody reactivity:
- Target protein:Nerve growth factor receptor
- Description:Human Nerve growth factor receptor(NGFR) ELISA Kit
- Size:96 wells
- Environmentally Preferable:
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf Life:6 months (4 °C); 12 months (−20 °C)
- Detection Range:1.56 - 100 ng/ml
- Storage Temperature:2…8 °C
- Application:Neuroscience
- Sample Volume:50 - 100 µl
- Sensitivity:0.2 ng/ml
- Regulatory Status:RUO
- Cat. No.:76715-962
- Detection wavelength: 450 nm