Rat Brain Derived Neurotrophic Facor(BDNF) ELISA Kit
Catalog # 76700-906
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Rat
- Target protein:Brain Derived Neurotrophic Facor
- Description:Rat Brain Derived Neurotrophic Facor(BDNF) ELISA kit
- Size:96 wells
- Sample Type:Serum, plasma, urine,tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:31.2 - 2000 pg/ml
- Storage Temperature:2…8 °C
- Sample Volume:50 - 100 μl
- Sensitivity:7.5 pg/ml
- Regulatory Status:RUO
- Cat. No.:76700-906
Specifications
About this item
- Detection wavelength: 450 nm