Goat Transforming Growth Factor β1,TGF-β1 ELISA Kit
Catalog # 76707-704
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Goat
- Target protein:Transforming Growth factor β1,TGF-β1
- Description:Goat Transforming Growth factor β1,TGF-β1 ELISA kit
- Size:96 wells
- Sample Type:Serum, plasma, urine,tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Storage Temperature:2…8 °C
- Sample Volume:50 - 100 μl
- Sensitivity:1 - 5 h
- Regulatory Status:RUO
- Cat. No.:76707-704
Specifications
About this item
- Detection wavelength: 450 nm