ContactUsLinkComponent
Wash Bottle for Water
Wash Bottle for Water
Catalog # 470344-058
Supplier:  Wards
undefined
Wash Bottle for Water
Catalog # 470344-058
Supplier:  Wards
Supplier Number:  CHWB1020A
Specifications
  • Capacity:
    500 ml
  • Description:
    Wash bottle for water
  • Cat. No.:
    470344-058
Specifications
About this item
Wash bottle for water with color-coded labels, wide mouth, self-venting design. Bottle is made of durable polypropylene, ideal for laboratory use with easy and precise dispensing.
  • Easy to read labeling, including chemical information and precautionary statements
  • Blue cap with built-in nozzle
  • Wide mouth for easy filling
  • Self venting design to eliminate leaks and dripping
  • Easy to control squeeze for dispensing
Made of low density polyethylene (LDPE) for superior quality and strength. Innovative flex ink won't fade, crack, or peel. Bottle is labeled for water and features chemical information, as well as safe handling and precautionary statements. Wide mouth for easy refilling. Each bottle features a two-piece construction, consisting of the bottle and leak-proof cap with built-in nozzle. Self venting design eliminates leaks and dripping. The low density polyethylene plastic is very responsive, with smooth bounce back that won't get stiff over time.
Each bottle measures 9" in total height (5.25" bottle alone) and 2.9" in diameter. Bottle opening has an inner diameter of 1.7" and an outer diameter measuring 1.9".