Organic Chemistry and Distillation 9 Piece Set
Catalog # 470343-654
Supplier: Wards
Specifications
- Description:Organic chemistry and distillation 9 piece set
- Environmentally Preferable:
- Cat. No.:470343-654
Specifications
About this item
Combining essential organic chemistry and distillation components, this nine-piece set offers versatility and convenience in laboratory work, all crafted from high-quality borosilicate glass for durability and optimal performance.
- Every piece of glassware you need to do basic organic chemistry and distillation
- Made of Borosilicate 3.3 glass
- All glassware is jointed (14/23)
- Made for preparations up to 30 to 40 g
Nine-piece set includes: (1) dropping funnel - 100 ml, (1) air leak/steam inlet tube, (1) Liebig condenser, (1) receiver adapter, (1) stopper, (1) flask, round bottom - 250 ml, (1) cone/screwcap adapter, (1) still head, (1) flask, pear shaped - 100 ml, (1) filter funnel - 24 mm. diameter perforated. Eisco borosilicate glass has the following heat tolerances: Strain point 515 °C, annealing point 565 °C, softening point 820 °C.