Specifications
- Description:22-piece organic chemistry set
- Environmentally Preferable:
- Cat. No.:470345-590
Specifications
About this item
This comprehensive organic chemistry set includes 22 items with a 10/19 joint size, offering versatility and convenience for various experiments, all made of durable borosilicate glass for reliable performance.
- This set contains 22 items with 10/19 ground joints for maximum versatility
- Glassware pieces are made of high quality borosilicate glass
- Over 20 standard assemblies can be constructed from simple reflux to fractional distillation
- Set 10 BU. Housed in die cut foam lined box
Stored in die cut foam lined box. Kit consists of: Consists of 1 Liebig condenser, 1 dropping funnel (20 ml), 1 pear shape flask (25 ml), 1 three neck flask (round bottom, 25 ml), 3 flasks (round bottom, 5 ml), 1 air leak/steam inlet, 1 thermometer (−10 to +250 °C), 1 dropping pipette, 1 rubber teat, 1 receiver adapter, 2 stoppers, 1 still head, 1 air condenser, 1 filter funnel, 1 receiver tube, 1 Willstatter Nail, 1 gas inlet tube, 1 drying tube, 1 stirrer. Autoclavable, with superior chemical durability. Low temperature gradient- can withstand high temperatures and thermal shocks. Eisco Borosilicate glass has the following heat tolerances: strain point 515 °C, annealing point 565 °C, softening point 820 °C.