ContactUsLinkComponent
2-Prong Double Adjustable Universal Clamp
2-Prong Double Adjustable Universal Clamp
Catalog # 470345-124
Supplier:  Wards
undefined
2-Prong Double Adjustable Universal Clamp
Catalog # 470345-124
Supplier:  Wards
Supplier Number:  CH0675B
Specifications
  • Description:
    2-prong double adjustable universal clamp
  • Cat. No.:
    470345-124
Specifications
About this item
Double adjustable universal clamp with 2 prong, integral boss head, and non-slip vinyl-coated rubber jaws, providing a secure grip and versatility.
  • This clamp with boss head is ideal for use with straight sides glassware apparatus such as tubes, flask necks, and burettes
  • Accommodates objects up to 3" and has non-slip vinyl coated rubber prongs
  • Adjustable thumb screws on both sides allows for a wider range of accommodation
  • 8" length, 4" width, 1" boss head opening
The item has an adjustable length of 7.5 to 8" and has a width of 4" at the thumb screws. The jaws measure 2" long and open 3" wide, and the jaws are adjustable on both sides with thumb screws.