Order Entry
United States
ContactUsLinkComponent
Plastic Double Burette Clamp
Plastic Double Burette Clamp
Catalog # 470344-856
Supplier:  Wards
undefined
Plastic Double Burette Clamp
Catalog # 470344-856
Supplier:  Wards
Supplier Number:  CH0262B

Specifications

  • Description:
    Plastic double burette clamp
  • Cat. No.:
    470344-856

Specifications

About this item

Double burette clamp made of polypropylene plastic, suitable for 10 mm or 12.5 mm rods.

  • Double clamp - Fits 2× rods 10 to 15 mm in diameter
  • Features two tapered wedges and a spring-loaded jack
  • Made of high quality polypropylene

Polypropylene double burette clamps come with easy-to-change wedges that hold rods of 10 mm or 12.5 mm diameter. Rugged yet lightweight, they are equipped with a spring-loaded jack that locks fir mly onto burettes to prevent slip. Non-metal build reduces the chance for accidental impact breaks of glass burettes.