Dog Granulocyte-Macrophage Colony Stimulating Factor,GM-CSF ELISA Kit
76703-144
:
- Assay Type:Sandwich
- Host:Dog
- Target protein:Granulocyte-Macrophage Colony Stimulating Factor,GM-CSF
- Description:Dog Granulocyte-Macrophage Colony Stimulating Factor,GM-CSF ELISA kit
- Size:96 wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf Life:6 months (4 °C); 12 months (−20 °C)
- Detection Range:Request Information
- Storage Temperature:2…8 °C
- Application:Immunology
- Sample Volume:50 - 100 μl
- Regulatory Status:RUO
- Cat. No.:76703-144
- Detection wavelength: 450 nm