Order Entry
United States
ContactUsLinkComponent
Mouse AMP Deaminase 2(AMPD2) ELISA Kit
Mouse AMP Deaminase 2(AMPD2) ELISA Kit
Catalog # 76705-794
Supplier:  AFG Bioscience
CAS Number:  
undefined
Mouse AMP Deaminase 2(AMPD2) ELISA Kit
Catalog # 76705-794
Supplier:  AFG Bioscience
Supplier Number:  EK732067
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Host:
    Mouse
  • Target protein:
    AMP deaminase 2
  • Description:
    Mouse AMP deaminase 2(AMPD2) ELISA kit
  • Size:
    96 wells
  • Sample Type:
    Serum, plasma, urine, tissue homogenates, cell culture supernates
  • Time to Results:
    1 - 5 h
  • Assay Principle:
    Quantitative
  • Shelf Life:
    6 months (4 °C); 12 months (−20 °C)
  • Detection Range:
    62.5 - 2000 pg/ml
  • Storage Temperature:
    2…8 °C
  • Sample Volume:
    50 - 100 μl
  • Sensitivity:
    7 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76705-794

Specifications

About this item

  • Detection wavelength: 450 nm