Mouse Transforming Growth Factor beta1(TGF-beta1) ELISA Kit
Catalog # 76715-630
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Mouse
- Target protein:Transforming Growth factor beta1
- Description:Mouse Transforming Growth factor beta1(TGF-beta1) ELISA kit
- Size:96 wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf Life:6 months (4 °C); 12 months (−20 °C)
- Detection Range:3.3 - 200 pg/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction
- Sample Volume:50 - 100 μl
- Sensitivity:0.6 pg/ml
- Regulatory Status:RUO
- Cat. No.:76715-630
Specifications
About this item
- Detection wavelength: 450 nm