ContactUsLinkComponent
Support Rod Clamp
Support Rod Clamp
Catalog # 470343-888
Supplier:  Wards
undefined
Support Rod Clamp
Catalog # 470343-888
Supplier:  Wards
Supplier Number:  CH0688G
Specifications
  • Description:
    Support rod clamp
  • Cat. No.:
    470343-888
Specifications
About this item
This adjustable clamp accommodates 10×1.5 threaded rods up to 12.5 mm and offers easy thumbscrew adjustment for secure attachment to table edges.
  • High performance, die casted metal support rod clamp. Tapped hole accommodates 10×1.5 threaded rods measuring up to 12.5 mm in diameter
  • Thumbscrew allows for an adjustable grip. The clamp fits table edges and surfaces up to 2.5" thick
  • Designed for use in professional laboratories, but is an ideal choice for science classrooms
Designed to securely fit table edges up to 2.5" thick with the adjustable thumbscrew. Stable and durable design. Clamp measures 6.25" in length and is 4" in width.