3 Prong Double Adjustable Universal Clamp
Catalog # 470343-638
Supplier: Wards
Specifications
- Description:3 prong double adjustable universal clamp
- Cat. No.:470343-638
Specifications
About this item
Double adjustable universal clamp with 3 prongs and non-slip vinyl-coated rubber jaws, offering a secure grip and versatility.
- 3 Prong Adjustable Universal Clamp with a 10" overall length and 0.375 diameter rod
- Perfect for holding both regular and irregular shaped glassware
- Accommodates objects up to 2.5"
- Adjustable thumb screws on both sides allows for a wider range of accommodation
- Non-slip vinyl coated rubber prongs allow for maximum grip and traction
This 3 prong clamp is perfect for holding both regular and irregular shaped glassware up to 2.5". It features non-slip vinyl coated rubber prongs for maximum grip and adjustable thumb screws on both sides for a wider range of accommodation.