ContactUsLinkComponent
Pipette Filler
Pipette Filler
Catalog # 470345-384
Supplier:  Wards
undefined
Pipette Filler
Catalog # 470345-384
Supplier:  Wards
Supplier Number:  CH0580D
Specifications
  • Description:
    Pipette filler
  • Cat. No.:
    470345-384
Specifications
About this item
Silicon bulb pipette filler with a 54 mm diameter and three pinch valves, suitable for handling noxious solutions and providing precise and controlled liquid dispensing.
  • Eisco Labs Silicon bulb pipette filler
  • For pipetting noxious solutions, microbial suspensions and other harmful and unpleasant solutions
  • Comprises a bulb and three pinch valves controlling influx and efflux of pipette contents
  • Bulb diameter is 54 mm
Silicone bulb pipette filler, for pipetting noxious solutions, microbial suspensions and other harmful solutions. Comprises a bulb and three pinch valves controlling influx and efflux of pipette contents.