Order Entry
United States
ContactUsLinkComponent
Double Burette Clamp
Double Burette Clamp
Catalog # 470344-440
Supplier:  Wards
undefined
Double Burette Clamp
Catalog # 470344-440
Supplier:  Wards
Supplier Number:  CH0256B

Specifications

  • Description:
    Double burette clamp
  • Cat. No.:
    470344-440

Specifications

About this item

Double burette clamp made of casted material.

  • Casted, with built-in boss head
  • Approximately 3.75" tall, 8" wide and 2.75" deep
  • To secure single burette
  • Durable, nonslip
  • Basic essential for classroom and laboratories

Secures up to two burettes. Fits burette up to 25 mm.