Order Entry
United States
ContactUsLinkComponent
Synthware Manifold, Solvent Transfer
Synthware Manifold, Solvent Transfer
Catalog # 60004-598
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Manifold, Solvent Transfer
Catalog # 60004-598
Supplier:  Kemtech America
Supplier Number:  M164380
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Height:
    140 mm
  • Cat. No.:
    60004-598
  • Joint size:
    24/40
  • Width:
    380 mm
  • Supplier no.:
    M164380

Specifications

About this item

Supplied with a high vacuum valve with a PTFE O-ring at the tip and PTFE wiper to protect Viton® O-ring on the shaft.

  • Heavy wall construction
  • 33 expansion borosilicate glass
  • Standard ground glass joints work with other brands

For purifying and transfering solvents under inert conditions.