Human Recombinant IL23 Protein (His & AVI Tag) (from HEK293 Cells), Biotinylated
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the human IL23 complex composed of human IL12B (NP_002178.2) (Met1-Ser328) and human IL23A (NP_057668.1) (Arg20-Pro189) linked by a polypeptide linker was fused with a c-terminal AVI tagged polyhistidine tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
- Shipping temperature: Dry ice
Specifications
- Conjugation:Biotin
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:−20…−80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method
- Biological Activity:Immobilized Human IL23 Receptor Protein (ECD, hFc Tag) (Cat:13840-H02H) at 2 μg/ml (100 μl/well) can bind Human IL-23 Protein (His and AVI Tag), Biotinylated (Cat:CT048-H27H-B), the EC50 is 80-480 ng/ml.
- Reconstitution Instructions:A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:IL12B & IL23A
- Protein/Peptide Name:IL12B & IL23A
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:The recombinant human IL23 complex (IL23A and IL12B) consists of 517 amino acids and predicts a molecular mass of 57.6 kDa.
- Sequence:Met1-Ser328 and Arg20-Pro189
- Formulation:Supplied as sterile 50 mM Tris, 100 mM Nacl, 10% glycerol, 0.02% Tween 80, 10% Trehalose, pH 8.5
Specifications
Frequently Bought Together