Mouse Recombinant LAG3/CD223/Lymphocyte activation gene 3 Protein (ECD) (from HEK293 Cells)
A DNA sequence encoding the mouse LAG3 (NP_032505.1) (Met1-Leu442) was expressed.
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:−20…−80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method
- Biological Activity:Measured by its ability to bind FGL1 Protein, Mouse, Recombinant (hFc Tag)(Cat:5A2001-M02H) in functional ELISA.
- Reconstitution Instructions:A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Ly66|CD223|LAG-3
- Protein/Peptide Name:LAG3
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:The recombinant mouse LAG3 consists of 419 amino acids and predicts a molecular mass of 45.25 kDa.
- Sequence:Met1-Leu442
- Formulation:Lyophilized from sterile PBS, pH 7.4
- Shipping Temperature:Ambient
Frequently Bought Together