Synthware Adapters, #15 O-Ring Joint to Inner Joint
Supplier: Kemtech America
Certificates
About this item
Adapter, with no. 15 O-ring joint to 14/20, 19/22, or 24/40 inner joint.
- Heavy wall construction
- 33 expansion Borosilicate glass
- Standard ground glass joints work with other brands
Designed for use with #15 O-Ring joint manifolds.
Frequently Bought Together