Anti-MPXV-A29L Mouse Monoclonal Antibody [clone: 32]
Catalog # 76628-280
Supplier: Sino Biological
Specifications
- Antibody Type:Primary
- Antigen Name:Monkeypox virus A29L protein
- Antigen Symbol:MPXV-A29L
- Clonality:Monoclonal
- Clone:0032
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Monkeypox virus (MPXV)
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Storage Buffer:0.2 μm filtered solution in PBS
- Storage Temperature:This antibody can be stored at 2...8 ℃ for one month without detectable loss of activity. Antibody products are stable for twelve months from date of receipt when stored at –20...–80 ℃. Preservative-Free. Avoid repeated freeze-thaw cycles.
- Shipping Temperature:Ambient
- Cat. No.:76628-280
Specifications
About this item
This product is a recombinant monoclonal antibody expressed from HEK293 cells.
Type: Primary
Antigen: MPXV-A29L
Clonality: Monoclonal
Clone: 32
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: MPXV