Anti-SIRPA Mouse Monoclonal Antibody [clone: 3]
Catalog # 76643-988
Supplier: Sino Biological
Specifications
- Antibody Type:Primary
- Antigen Name:signal regulatory protein alpha
- Antigen Symbol:SIRPA
- Clonality:Monoclonal
- Clone:03
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:PTPNS1|CD172A|MYD-1|SHPS1|P84|SIRP alpha|BIT|SIRP|MFR
- Storage Buffer:0.2 μm filtered solution in PBS
- Storage Temperature:This antibody can be stored at 2...8 ℃ for one month without detectable loss of activity. Antibody products are stable for twelve months from date of receipt when stored at –20...–80 ℃. Preservative-Free. Avoid repeated freeze-thaw cycles.
- Shipping Temperature:Ambient
- Cat. No.:76643-988
Specifications
About this item
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SIRPA (rh SIRPA; Catalog No: 11612-H08H; NP_542970.1; Met1-Arg370, Gly 75 Ala). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Type: Primary
Antigen: SIRPA
Clonality: Monoclonal
Clone: 3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human