Anti-HGF Rabbit Polyclonal Antibody
Catalog # 76643-880
Supplier: Sino Biological
Specifications
- Antibody Type:Primary
- Antigen Name:hepatocyte growth factor
- Antigen Symbol:HGF
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:HPTA|DFNB39|HGFB|Hepatocyte Growth Factor|SF|F-TCF|F-TCFB
- Storage Buffer:0.2 μm filtered solution in PBS
- Storage Temperature:This antibody can be stored at 2...8 ℃ for one month without detectable loss of activity. Antibody products are stable for twelve months from date of receipt when stored at –20...–80 ℃. Preservative-Free. Avoid repeated freeze-thaw cycles.
- Shipping Temperature:Ambient
- Cat. No.:76643-880
Specifications
About this item
Produced in rabbits immunized with purified, recombinant Human HGF/Hepatocyte Growth Factor (rh HGF/Hepatocyte Growth Factor; Catalog No: 10463-HNAS; NP_000592.3; Met1-Ser728). HGF/Hepatocyte Growth Factor specific IgG was purified by Human HGF/Hepatocyte Growth Factor affinity chromatography.
Type: Primary
Antigen: HGF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human