Anti-NP-CoV Mouse Monoclonal Antibody [clone: 124]
76632-684
- Antibody Type:Primary
- Antigen Name:Coronavirus Nucleocapsid
- Antigen Symbol:NP-CoV
- Clonality:Monoclonal
- Clone:124
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:SARS-CoV-2
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:Nucleoprotein|coronavirus Nucleocapsid|cov np|ncov NP|coronavirus NP|Nucleocapsid|NP|coronavirus Nucleoprotein|novel coronavirus Nucleoprotein
- Storage Buffer:0.2 μm filtered solution in PBS
- Storage Temperature:This antibody can be stored at 2...8 ℃ for one month without detectable loss of activity. Antibody products are stable for twelve months from date of receipt when stored at −20...−80 ℃. Preservative-Free. Avoid repeated freeze-thaw cycles.
- Shipping Temperature:Ambient
- Cat. No.:76632-684
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, SARS-CoV-2 (2019-nCoV) Nucleocapsid (Catalog No: 40588-V08B; YP_009724397.2(335Gly/Ala); Met1-Ala419). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Type: Primary
Antigen: NP-CoV
Clonality: Monoclonal
Clone: 124
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: SARS-CoV-2