Anti-CDON Rabbit Polyclonal Antibody
Catalog # 76643-656
Supplier: Sino Biological
Specifications
- Antibody Type:Primary
- Antigen Name:cell adhesion associated, oncogene regulated
- Antigen Symbol:CDON
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:HPE11|ORCAM|CDO|CDON1
- Storage Buffer:PBS, pH 7.0 with 0.03% Proclin300
- Storage Temperature:This antibody can be stored at 2...8 ℃ for one month without detectable loss of activity. Antibody products are stable for twelve months from date of receipt when stored at −20...−80 ℃. Preservative-Free. Avoid repeated freeze-thaw cycles.
- Shipping Temperature:Ambient
- Cat. No.:76643-656
Specifications
About this item
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CDO/CDON, and purified by antigen affinity chromatography.
Type: Primary
Antigen: CDON
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human