Anti-LAYN Mouse Monoclonal Antibody [clone: 3F7D7E2] (Biotin)
76644-434
- Antibody Type:Primary
- Antigen Name:layilin
- Antigen Symbol:LAYN
- Clonality:Monoclonal
- Clone:3F7D7E2
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Layilin
- Storage Buffer:0.2 μm filtered solution in PBS
- Storage Temperature:This antibody can be stored at 2...8 ℃ for one month without detectable loss of activity. Antibody products are stable for twelve months from date of receipt when stored at −20...−80 ℃. Preservative-Free. Avoid repeated freeze-thaw cycles.
- Shipping Temperature:Ambient
- Cat. No.:76644-434
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Layilin (rhLayilin; Catalog No: 10208-H08H; Met1-Glu220;NP_849156.1; NP_849156.1) and then biotinylated. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and then biotinylated.
Type: Primary
Antigen: LAYN
Clonality: Monoclonal
Clone: 3F7D7E2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human