Anti-DPP6 Rabbit Polyclonal Antibody
Catalog # 76644-368
Supplier: Sino Biological
Specifications
- Antibody Type:Primary
- Antigen Name:dipeptidyl peptidase like 6
- Antigen Symbol:DPP6
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Storage Buffer:0.2 μm filtered solution in PBS
- Storage Temperature:This antibody can be stored at 2...8 ℃ for one month without detectable loss of activity. Antibody products are stable for twelve months from date of receipt when stored at −20...−80 ℃. Preservative-Free. Avoid repeated freeze-thaw cycles.
- Shipping Temperature:Ambient
- Cat. No.:76644-368
Specifications
About this item
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human DPP6, and purified by antigen affinity chromatography.
Type: Primary
Antigen: DPP6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human