Magid® T-REX® Flex Series® Knit Impact Gloves with NitriX Grip Palm, Cut Level A9
The Magid® T-REX® Flex Series® TRX795 NitriX® Grip Technology Palm Impact Glove delivers high quality A9 cut resistance and Impact Level 1 protection at a great value.
- 13-gauge Hyperon® shell delivers breathability, comfort, and cut protection
- Nitrile reinforced thumb saddle enhances durability at a critical wear point
- Elastic knit wrist cuff ensures a secure fit
- Machine washable for longer service life
- ANSI Level A9 Cut Resistance, ANSI Level 5 Abrasion Resistance and ANSI Level 4 Puncture Resistance
- ANSI/ISEA 138 Impact Level 1 (Level 1 protection on knuckles and Level 1 protection on fingers)
NitriX® Grip Technology palm coating delivers rock-solid grip in oily applications. M-Force Defense System® back-of-hand protection extends all the way to the finger-tip and between thumb and index finger to deflect and absorb impacts.
- Catalog No:
- 76635-844
- 76635-836
- 76635-834
- 76635-838
- 76635-840
- 76635-842
- Ambidextrous/Hand-specific:
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Disposable/Reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Powdered/Powder-free:
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Gauge:
- 13
- 13
- 13
- 13
- 13
- 13
Frequently Bought Together